Learn More
ATCC Pneumocystis carinii
Supplier:  ATCC PRA159
Encompass
Pneumocystis carinii strain M167-6 was isolated in October 2003 from the lungs of an immunosuppressed Lewis male rat in Cincinnati, Ohio. This parasitic protozoan is cultivated in rats and has applications in opportunistic pathogen research.
Catalog No. 50-238-2654