Learn More
ABclonal Technology PNLIPRP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A12895100UL
Encompass_Preferred
This gene encodes a lipase that hydrolyzes galactolipids, the main components of plant membrane lipids. An allelic polymorphism in this gene results in both coding and non-coding variants, the reference genome represents the non-coding allele.
Catalog No. 50-001-15411