Learn More
Aviva Systems Biology Corp PNPLA1 Antibody - middle region (ARP49112_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49112_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNPLA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PNPLA1. Alternate names: FLJ38755, MGC119319, MGC119320, MGC119321, dJ50J22.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62963