Learn More
Apexbio Technology LLC PNPP 4264-83-9 1g
SDP
Supplier:  Apexbio Technology LLC C69591000
Encompass_Preferred
PNPP (4264-83-9) is a chromogenic non-proteinaceous substrate utilized to assess phosphatase activity It is designed to serve as an analytical reagent for quantifying the activity of alkaline or acid phosphatases through enzymatic hydrolysis resulting in the formation of para-nitrophenol a yellow product with measurable absorbance PNPP exerts its application primarily through substrate conversion upon phosphatase-mediated hydrolysis Based on these properties PNPP holds research potential in enzymatic assays enzyme kinetics studies and the investigation of signal transduction pathways and enzyme-mediated biological processes in biomedical laboratories
Catalog No. 50-000-90787