Learn More
Surmodics Inc AP Yellow One Component Microwell Substrate (pNPP) with Stabilizer Pellets, 1L
SDP
Supplier:  Surmodics Inc PNPS100001
Encompass_Preferred
The formulation consists of p-nitrophenylphosphate (p-NPP) solution with stabilizing pellets to for long term stability. The product is stable for 3 years from the date of manufacture.
Catalog No. NC1517289