Learn More
Bioworld pNPP (p-Nitrophenyl Phosphate), Disodium, hexahydrate, 1 g
Supplier:  Bioworld 215301101
Encompass_Preferred
Colorimetric alkaline phosphatase soluble substrate, p-Nitrophenyl Phosphate (pNPP) is the substrate of choice for use with alkaline phosphatase in Enzyme Linked Immunosorbant Assay (ELISA) procedures. The reaction produces a soluble end product that is yellow in color. Can be used to perform kinetic and endpoint determinations. Also used as a phosphorylating agent in nucleotide synthesis.Specifications:CAS Number: 4264-83-9Molecular Weight: 371.14Molecular Formula: C6H4NO6P•2Na•6H2OPhysical Appearance: White to yellow crystalline powderStorage:Store at 4° C, Unstable in solution; reconstitute just prior to use.
Catalog No. 50-198-938