Learn More
Cayman Chemical Pntoprazole N-oxIde 50mg
SDP
Supplier:  Cayman Chemical 3544950 MG
Encompass_Preferred
A potential impurity found in bulk preparations of pantoprazole
Catalog No. 50-290-9602