Learn More
Cayman Chemical Pntoprazole sulfon 5mg
SDP
Supplier:  Cayman Chemical 222525 MG
Encompass_Preferred
A pantoprazole metabolite; formed by CYP2C19 and CYP3A4 metabolism of pantoprazole
Catalog No. 50-298-6183