Learn More
Cayman Chemical PNU 112455A 25mg
SDP
Supplier:  Cayman Chemical 3343825 MG
Encompass_Preferred
A Cdk2 and Cdk5 inhibitor (Ki = 2 µM for both); inhibits dexamethasone-induced hyperphosphorylation of tau and cell death in PC12 cells expressing human tau at 1 and 10 µM
Catalog No. 50-294-6722