Learn More
Medchemexpress LLC Podoplanin protein, rat (HEK293, hFc) | >95.0% | 5 UG

Supplier: Medchemexpress LLC HYP746245UG
Recombinant rat podoplanin extracellular domain expressed in HEK293 cells and supplied as a C-terminal human Fc fusion. Provided at research-grade purity for biochemical and cell-based assays, the product is intended for use in ligand-receptor interaction studies, platelet activation assays, and cell adhesion or migration experiments. Technical documents such as data sheet, SDS, and COA are available from the manufacturer.
- Recombinant extracellular domain expressed in HEK293 cells.
- C-terminal human Fc fusion for increased stability and detection.
- High purity as determined by reducing SDS-PAGE (>95%).
- Supplied in microgram quantities suitable for in vitro assays.
- Sequence and technical documents available (data sheet, SDS, COA).
- Suitable for binding, platelet activation, and migration studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.