Learn More
ABclonal Technology POFUT2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A122231000UL
Encompass_Preferred
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor (EGF, MIM 131530)-like repeats or thrombospondin (THBS, see MIM 188060) type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates (Luo et al., 2006 [PubMed 16464857]).
Catalog No. 50-001-14943