Learn More
Medchemexpress LLC POL1-IN-1 | 1822358-25-7 | 98.1% | 356.42 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112062
Encompass_Preferred
POL1-IN-1 is an RNA polymerase 1 (POL1) inhibitor with an IC₅₀ of less than 0.5 μM. It inhibits ribosome biogenesis by preventing POL1 transcription.
  • RNA polymerase 1 (POL1) inhibitor
  • Inhibits ribosome biogenesis
  • Used for research purposes
Catalog No. 50-004-39048