Learn More
WELCH MATERIALS INC Ultisil Polar RP, 10µm, 30×250mm
Supplier:  WELCH MATERIALS INC H0262131108
Encompass_Preferred
Prep Column, Ultisil Polar RP, 10µm, 30×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, L60, Carbon Loading: 18%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052699
Only null left
Add to Cart
Request a Quote