Learn More
WELCH MATERIALS INC Ultisil Polar RP, 10µm, 120Å, 4.6×250mm
Supplier:  WELCH MATERIALS INC H0021541043
Encompass_Preferred
Analytical Column, Ultisil Polar RP, 10μm, 120Å, 4.6×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, L60, Carbon Loading: 18%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052224