Learn More
Aviva Systems Biology Corp POLD3 Antibody - C-terminal region (ARP62093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLD3. It was validated on Western Blot. . Alternate names: KIAA0039, MGC119642, MGC119643, P66, P68ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-565