Learn More
*CARBOMER INC Poloxamer 237 NF, 9003-11-6, MFCD00082049, 250 g
Supplier:  *CARBOMER INC 801153250G
Encompass
Molecular Formula: HO((C2H4O)m(C3H6O)n(C2H4O)m)H, Relative Molar Mass: Mw 6,840-8,830, Synonym: Poly(ethylene)mpoly(propylene)n glycol block copolymer m = 64 n = 37.
Catalog No. NC1527181