Learn More
Medchemexpress LLC HY-77195 5mg Medchemexpress, Poloxime CAS:17302-61-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY771955MG
Encompass_Preferred
Medchemexpress, HY-77195 5mg Poloxime CAS:17302-61-3 Poloxime, a hydrolysis product of poloxin, is a non-ATP-competitive Plk1 inhibitor, with moderate Plk1 inhibitory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2286