Learn More
Apexbio Technology LLC Poloxin 321688-88-4 50mg
SDP
Supplier:  Apexbio Technology LLC A373250
Encompass_Preferred
Poloxin (CAS 321688-88-4) is a synthetic small molecule that selectively inhibits the polo-box domain (PBD) of Polo-like kinase 1 (Plk1) a serine/threonine-protein kinase involved in cell division By targeting the Plk1 PBD Poloxin interferes with substrate recognition and localization of Plk1 disrupting key mitotic processes This compound is utilized in biomedical research to investigate Plk1-mediated signaling pathways and to explore the biological consequences of PBD inhibition in cancer cell models contributing to the understanding of mitotic regulation and the development of potential anticancer strategies
Catalog No. 50-000-96468