Learn More
ABclonal Technology POLR2H Rabbit pAb
SDP
Supplier:  ABclonal Technology A1333920UL
Encompass_Preferred
The three eukaryotic RNA polymerases are complex multisubunit enzymes that play a central role in the transcription of nuclear genes. This gene encodes an essential and highly conserved subunit of RNA polymerase II that is shared by the other two eukaryotic DNA-directed RNA polymerases, I and III. Alternative splicing results in multiple transcript variants of this gene.
Catalog No. 50-000-97512