Learn More
Medchemexpress LLC Poly(2-hydroxyethyl methacrylate) | 25249-16-5 | 300000 (Average) | 10 G
SDP
Supplier:  Medchemexpress LLC HY112531B10G
Encompass_Preferred
Poly(2-hydroxyethyl methacrylate) is a biocompatible, optically transparent, hydrophilic, and non-degradable polymer known for excellent cytocompatibility and minimal immunological response. It is suitable for various biomedical applications, including bone tissue regeneration, wound dressings, and drug delivery.
  • Forms hydrogels by absorbing water or biological fluids through hydrophilic pendant groups.
  • Utilized in applications such as soft contact lenses, tissue regeneration, cancer research, ocular drug delivery, artificial skin, and artificial cornea.
  • Can be loaded with Quercetin (QT) to synthesize QT-pHEMA-GelCryoMem, demonstrating antimicrobial activity against *S. aureus*.
  • Applicable in wound dressings, scaffolds, or drug delivery.
  • Can be used in the synthesis of spongy materials with antimicrobial properties, suitable for wound and burn dressing applications.
Catalog No. 50-004-36005