Learn More
Medchemexpress LLC Poly(2-hydroxyethyl methacrylate) (Mv 300000) | 25249-16-5 | 300000 (Average) | 1 G
SDP
Supplier:  Medchemexpress LLC HY112531B1G
Encompass_Preferred
Poly(2-hydroxyethyl methacrylate) (Mv 300000) is a biocompatible, optically transparent, hydrophilic, and non-degradable polymer with excellent cytocompatibility, eliciting a minimal immunological response from host tissue. It can be used for different biomedical applications, such as bone tissue regeneration, wound dressings, and drug delivery. It also absorbs water or biological fluids via its hydrophilic pendant groups to form a hydrogel.
  • Used for bone tissue regeneration, wound dressings, and drug delivery
  • Forms a hydrogel for applications like soft contact lenses, tissue regeneration, and cancer research
  • Can be loaded with Quercetin (QT) to synthesize QT-PHEMA-GelCryoMem for antimicrobial activity
  • Used in the synthesis of spongy materials with antimicrobial properties for wound and burn dressing applications
Catalog No. 50-004-38006