Learn More
Sigma Aldrich Fine Chemicals Biosciences Poly(2-hydroxyethyl methacrylate), 25249-16-5, MFCD00084374, 10 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P393210G
Encompass
Linear Formula: (C6H10O3)n, BioReagent, powder, suitable for cell culture, density: 1.15 g/mL at 25 °C, Synonym: Poly(2-HEMA), Poly-HEMA. ** For R&D use only. Not for drug, household, or other uses.
Catalog No. NC0215770