Learn More
Sino Biological Peptide Substrates: Poly (6:2:5:1 Ala,Glu,Lys,Tyr) Peptide
Supplier:  Sino Biological P6058
Encompass_Preferred
The Poly (6:2:5:1 Ala,Glu,Lys,Tyr) substrate peptide is used as universal substrate for protein tyrosine kinases.
Catalog No. 50-256-8155