Learn More
Supply Solutions Poly (A) Polyadenylic acid
Supplier:  Supply Solutions SIGMA 10108626001
Encompass
Poly (A) Polyadenylic acid. Working concentration: 0.5 mg/ml; Working solution: 0.5 mg/ml is recommended; Physical form: Lyophilizate, potassium salt; Application: Poly(A) has been used for droplet digital PCR (ddPCR) assay; General description: Poly(A) is used as a carrier for quantitative precipitation of DNA and RNA.
Catalog No. 50-100-4984