Learn More
Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | MFCD00084394 | (C3H4O2)n | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW243303O100MG
Encompass_Preferred
Poly(acrylic acid) (Mv 1,250,000) is a high-molecular-weight anionic polymer supplied as a viscous liquid for research use. It is used as a corrosion inhibitor, surface stabilizer, and biomaterial component in life-science and materials research. The viscous formulation supports coating, formulation, and analytical applications; follow supplier storage recommendations.
Catalog No. 50-002-80787