Learn More
Polyscience polyclean CLARIFIER
Supplier: Polyscience 004300040
PolyScience™ PolyScience° CLARIFIER 8oz BottleKeeps reservoirs clean and odor-free. Concentrated: 8 oz (237 ml) treats approximately 200 gallons (757 liters).