Learn More
Medchemexpress LLC Poly-D-lysine hydrobromide | 27964-99-4 | 84000 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139201B
Encompass_Preferred
Poly-D-lysine hydrobromide (MW 84000) is a synthetic polymeric substrate commonly used in neural cell culture. This compound also functions as a CaSR agonist peptide. Its polycationic nature allows it to stimulate the endocytosis of various cell types. It exhibits stability against pancreatic peptidases.
  • Synthetic polymeric substrate widely utilized in neural cell culture.
  • Acts as a CaSR agonist peptide.
  • Stimulates endocytosis in different cell types due to its polycationic properties.
  • Stable in the presence of pancreatic peptidases.
Catalog No. 50-003-48122