Learn More
Medchemexpress LLC Poly-D-lysine hydrobromide | 27964-99-4 | MFCD00131934 | 99.9% | 70,000-150,000 | (C6H14N2O2)n·xHBr | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1392015MG
Encompass_Preferred
Poly-D-lysine hydrobromide (MW 70,000-150,000) is a positively charged synthetic polyamino acid supplied as the hydrobromide salt. It is commonly used to coat cultureware to enhance adhesion of neuronal and other adherent mammalian cells. Supplied in small research quantities with supporting analytical and safety documentation.
  • Promotes cell adhesion on plastic and glass surfaces.
  • High purity suitable for cell-culture and biochemical assays.
  • Hydrobromide salt form, water-soluble for coating preparations.
  • Available in small research pack sizes for laboratory use.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-23419