Learn More
Medchemexpress LLC Poly-D-lysine hydrobromide (MW 1000-5000) | 27964-99-4 | 1000-5000 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13920150MG
Encompass_Preferred
Poly-D-lysine hydrobromide (MW 1000-5000), also known as PDLHB (MW 1000-5000), is a synthetic cationic polypeptide polymer. It is commonly used in various research applications.
  • Commonly used as a cell adhesion promoter
  • Commonly used as a surface coating molecule
  • Also known as a calcium-sensing receptor (CaSR) agonist peptide
Catalog No. 50-003-52002