Learn More
Apexbio Technology LLC Poly(I:C), 24939-03-5, MFCD00131957, 10 mg
SDP
Supplier:  Apexbio Technology LLC B555110
Encompass_Preferred
MF: C19H27N7O16P2, MW: 671.41, Solubility: 21.5mg/mL in H2O,insoluble in DMSO. Poly (I:C), a synthetic double-stranded RNA (dsRNA) analog, is an immunostimulant that acts as the most potent interferon (IFN) inducer.
Catalog No. 50-115-2136