Learn More
Medchemexpress LLC Poly-L-Glutamic acid (MW 700000) | 25513-46-6 | 98.4% | 700000 (Average) | 50 G
SDP
Supplier:  Medchemexpress LLC HY11211150G
Encompass_Preferred
Poly-L-Glutamic acid (MW 700000) is a compound carrier. It can be used for the targeted delivery of anticancer compounds.
  • Compound carrier
  • Can be used for the targeted delivery of anticancer compounds
Catalog No. 50-004-36026