Learn More
AdipoGen Poly(L-lactide) 1.0 dl/g,Chemical. CAS: 26161-42-2
SDP
Supplier:  AdipoGen CDXP0013G050
Encompass_Preferred
Poly(L-lactide) 1.0 dl/g, AdipoGen Life Sciences. Chemical. CAS: 26161-42-2. Formula: (C3H4O2)n. MW: 59,000-101,000. Synthetic. Polylactides (PLA) used in material sciences. Biodegradable Polymer. Lactide and Glycolide Polymers. 26161-42-2
Catalog No. 50-117-7662