Learn More
Sciencell Research Laboratories POLY-L-LYSINE 10MG/ML 1ML
Supplier: Sciencell Research Laboratories 0413
Poly-L-Lysine is a synthetic compound that is a highly positively charged amino acid chain. It is commonly used as a coating agent to promote cell adhesion and improve the survival of primary cells in culture.