Learn More
Electron Microscopy Sciences Poly-L-Lysine Coated Slides, 1" x 3", 144/box
SDP
Supplier:  Electron Microscopy Sciences 6341002
Encompass_Preferred
Our premium glass slides with a frosted end are now available pre-coated. Pre-coated with Poly-L-Lysine, these slides improve the adhesion of tissues to the slide.
Catalog No. 50-279-89