Learn More
Medchemexpress LLC Poly-L-lysine hydrobromide | 25988-63-0 | 1000-5000 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY126437I50MG
Encompass_Preferred
Poly-L-lysine hydrobromide (MW 1000-5000) is a low molecular weight polycationic peptide composed of L-lysine monomers. It can be utilized for gene delivery research in cancer, specifically in lung cancer and breast cancer, as well as in bone regeneration-related diseases.
  • Low molecular weight polycationic peptide composed of L-lysine monomers.
  • Can be used for gene delivery research in cancer (such as lung cancer, breast cancer).
  • Can be used for gene delivery research in bone regeneration-related diseases.
Catalog No. 50-004-38064