Learn More
Medchemexpress LLC Poly-L-lysine hydrochloride | 26124-78-7 | 30000-50000 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY126437A10MG
Encompass_Preferred
Poly-L-lysine hydrochloride is a nonspecific attachment factor for cells, useful in promoting cell adhesion to solid substrates. It enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface, and acts as a strong-attraction regulator. It is also an antibacterial cationic peptide. For research use only.
  • Promotes cell adhesion to solid substrates
  • Enhances electrostatic interaction for cell membrane and culture surface
  • Acts as a strong-attraction regulator
  • Promotes liquid-liquid phase separation (LLPS) at low concentrations
  • Suppresses LLPS at high concentrations
  • Functions as an antibacterial cationic peptide
Catalog No. 50-004-37824