Learn More
Medchemexpress LLC Poly-L-lysine (hydrochloride) | 26124-78-7 | 30000-50000 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY126437A50MG
Encompass_Preferred
Poly-L-lysine hydrochloride is a nonspecific attachment factor for cells. It is useful in promoting cell adhesion to solid substrates by enhancing the electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. It also acts as a strong-attraction regulator, promoting liquid-liquid phase separation (LLPS) at low concentrations and suppressing LLPS at high concentrations. This compound is an antibacterial cationic peptide.
  • Nonspecific attachment factor for cells
  • Promotes cell adhesion to solid substrates
  • Enhances electrostatic interaction between cell membrane ions and the culture surface
  • Promotes liquid-liquid phase separation (LLPS) at low concentrations
  • Suppresses liquid-liquid phase separation (LLPS) at high concentrations
  • Antibacterial cationic peptide
Catalog No. 50-004-35789