Learn More
Newcomer Supply Poly-L-Lysine Adhesive Coated Slides, White, Gross
SDP
Supplier:  Newcomer Supply 5010
Encompass
Excellent for difficult tissues, frozen sections and special stains (especially Silvers). Specifications: “Adhesive” imprinted below painted area, Size: 75 x 25mm, 1mm thick, Color: White, Unit: Approx. 144 slides/gross.
Catalog No. NC9951478