Learn More
Akina Inc Poly(lactide-co-glycolide)-b-poly(ethyelen glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Supplier:  Akina Inc AK1421G
Encompass
PLGA-PEG-PLGA (Mw ~1,500:1,500:1,500 Da) LA:GA 95:5 Poly(lactide-co-glycolide)-b-poly(ethyelen glycol)-b-poly(lactide-co-glycolide) (per g)
Catalog No. NC2229294