Learn More
Akina Inc Poly(lactide-co-glycolide)-b-Poly(ethylene glycol)-b-Poly(lactide-co-glycolide) (per g)
SDP
Supplier:  Akina Inc AK091
Encompass
PLGA-PEG-PLGA (Mw ~1,500:1,500:1,500 Da) LA:GA 6:1 Poly(lactide-co-glycolide)-b-Poly(ethylene glycol)-b-Poly(lactide-co-glycolide) (per g)
Catalog No. NC3845550