Learn More
Sigma Organic Chemistry Poly(methyl methacrylate) | 500G | 9011-14-7 | MFCD00134349
Supplier: Sigma Organic Chemistry 182230500G
Poly(methyl methacrylate), 500G
About This Item:
Density: 1.188 g/mL at 25 C
Refractive Index: n20/D 1.49
Storage: room temp
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.