Learn More
Medchemexpress LLC Poly(methyl vinyl ether-alt-maleic anhydride) | 9011-16-9 | MFCD00134013 | >98.0% | 216,000 (average) | (C4H2O3·C3H6O)n | 100 G
SDP
Supplier:  Medchemexpress LLC HYW392258100G
Encompass_Preferred
Poly(methyl vinyl ether-alt-maleic anhydride) is an alternating copolymer (average Mw 216,000) used in materials and biomedical research. According to the supplier datasheet, it can be used to prepare antibacterial hydrogels that show good mechanical strength and excellent swelling ability.
  • Alternating copolymer structure suitable for polymer-based formulations.
  • Average molecular weight of 216,000 (average).
  • Suitable for preparation of antibacterial hydrogels.
  • Provides good mechanical strength in hydrogel networks.
  • Exhibits excellent swelling ability for absorbent applications.
Catalog No. 50-002-85143