Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-[2-(ditetradecylamino)-2-oxoethyl]-ω-methoxy- | 1849616-42-7 | 98.0% | 2464.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138300
Encompass_Preferred
ALC-0159 is a polyethylene glycol (PEG) lipid conjugate that can be used as a vaccine excipient. It appears as a white to off-white solid.
  • Polyethylene glycol (PEG) lipid conjugate
  • Used as vaccine excipient
  • Applied in lipid nanoparticle (LNP) preparation
  • For research use only
Catalog No. 50-003-50470