Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl) α-(folate-conjugated DSPE) ammonium salt | 1236288-25-7 | 95.7% | C65H114N11O16P | 10MG
SDP
Supplier:  Medchemexpress LLC HY14400910MG
Encompass_Preferred
DSPE-PEG-Folate is a PEGylated phospholipid conjugated to folic acid used to confer folate-mediated targeting to liposomal and lipid nanoparticle formulations. It promotes receptor-mediated uptake by folate-receptor-expressing cells and supports targeted delivery and imaging applications.
  • Folate-mediated targeting to folate receptor-expressing cells.
  • Pegylation enhances circulation time and colloidal stability.
  • Compatible with liposome and lipid nanoparticle formulations.
  • High purity suitable for research and formulation work.
  • Available in small-scale sizes for development and testing.
Catalog No. 50-002-58293