Learn More
MSC Honeywell CoreShield 3.0 Cut Resistant Gloves, Nitrile, 18 Gauge, Size 8, 10 Pairs
Supplier: MSC 37100054
These cut-resistant gloves feature an 18 gauge seamlessly knitted liner made of recycled polyester and polyethylene, finished with a nitrile micro-foam coating for superior grip and durability. Engineered for high performance, they provide effective protection while maintaining dexterity and comfort.
- Cut level A2/B protection
- Lightweight, glass fiber-free construction
- Enhanced grip with nitrile micro-foam coating
- Integrated thumb reinforcement for added durability
- Silicone-free and antistatic properties
- Certified for food handling
- Breathable coating for increased comfort
- Washable for extended use
- Low DMF content (<100 ppm)
- Color-coded markings for easy identification