Learn More
MSC PIP Global Safety Cut Resistant Gloves, 18 Gauge, Nitrile Foam Coated, Size S, 100/Cs.
Supplier:  MSC 37100609
Encompass
These CoreShield 3.0 cut-resistant gloves feature an 18 gauge, fiberglass-free, recycled polyester and PE liner for lightweight protection and dexterity. Designed for industrial use, they provide reliable defense against cuts, abrasions, and punctures in environments ranging from automotive assembly to food processing.
  • ANSI cut level A2 and EN 388 4X42B protection
  • Nitrile micro-foam coating on palms and fingertips for superior grip
  • ESD safe and silicone-free construction
  • Reinforced thumb crotch for increased durability
  • Touchscreen compatible and reusable/washable design
  • Certified for food contact under FDA 21 CFR and EC 1935/2004
Catalog No. 50-005-08775