Learn More
MSC PIP CoreShield 3.0 Cut-Resistant Gloves, HPPE, 18 Gauge, A2, 10/Pk.
Supplier: MSC 65398661
These cut-resistant gloves feature an 18-gauge, fiber-free engineered yarn liner with a foam nitrile coating on the palm and fingers. Designed for tactile sensitivity and durability, they offer protection in industrial environments and are compatible with touchscreen devices. The seamless construction enhances comfort, while a reinforced thumb crotch improves longevity in high-stress areas.
- ANSI cut level A2 protection
- Breathable foam nitrile coating provides reliable grip in dry, wet, and oily conditions
- Touchscreen compatible for device operation
- Reinforced thumb crotch for extended glove life
- ESD and anti-static properties
- Food contact compliant per 21 CFR 177.2600 and EC 1935/2004
- Reusable and launderable design