Learn More
Bio X Cell InVivoMAb Polyclonal Rat IgG, Protein G Purified, ≥95% Purity
Supplier: Bio X Cell BE0094R025MG
This polyclonal rat IgG is purified from rat serum, offering an ideal non-reactive control for polyclonal rat IgG antibodies in various in vivo and in vitro applications. It is formulated in PBS, pH 7.0, without stabilizers or preservatives, ensuring high purity and reliability for research use.
- Isotype: Rat IgG
- Recommended for use with InVivoPure pH 7.0 Dilution Buffer
- Unconjugated
- Endotoxin levels ≤1 EU/mg
- Purity ≥95%, determined by SDS-PAGE
- Sterility assured by 0.2 μm filtration
- Purified using Protein G chromatography
- Molecular weight of 150 kDa
- Negative for murine pathogen tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.