Learn More
Vector Laboratories POLY RAB ANTI HU HPMS1/PMS1
SDP
Supplier:  Vector Laboratories LSC352675100
Encompass
PMS1 antibody LS-C352675 is an unconjugated rabbit polyclonal antibody to PMS1 (HPMS1) (Internal) from human. It is reactive with human, mouse and rat. Validated for IHC and WB.100UL; LIFESPAN BIOSCIENCES mfg# LS-C352675-100
Catalog No. NC1881359