Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Value Film Liner, 20 in x 2400 in, 1 Case

Supplier:  MSC 39573779

Encompass

This polyethylene plastic film liner provides reliable protection for equipment and machinery from moisture, dirt, and dust. It serves as an economical alternative to traditional drop cloths and is suitable for various applications, including use as a moisture barrier for walls or covering lumber on construction sites.

  • Clear polyethylene material
  • Thickness of 2 mil
  • Width of 20 in
  • Length of 2400 in
  • Packaged in a case

Catalog No. 50-191-7796


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.